Learn More
Cayman Chemical SynthetIc NIgerose 25mg
SDP
Supplier:  Cayman Chemical 3741925 MG
Encompass_Preferred
A disaccharide; prevents increases in MDA and ROS levels and inhibits LDH release in HL-60 cells post-cryopreservation at 150 µM in combination with salidroside
Catalog No. 50-291-5780