Learn More
Cayman Chemical Nimbin, Purity: ≥98%, CAS: 5945-86-8, 5 mg
SDP
Catalog No. 502991640
Encompass_Preferred
A limonoid with diverse biological activities; inhibits the growth of D. oryzae by 64.8% at 1,000 ppm; has antifeedant activity against S. littoralis in a choice test at 0.02-2,000 μM; decreases melanin content to 20.9% of control in B16 murine melanoma cells at 25 μg/ml; inhibits TPA-induced activation of EBV-EA in Raji cells (IC50 = 499 mol ratio/32 pmol TPA)
Catalog No. 50-299-1640 Supplier Cayman Chemical Supplier No. 298845 MG
A limonoid with diverse biological activities; inhibits the growth of D. oryzae by 64.8% at 1,000 ppm; has antifeedant activity against S. littoralis in a choice test at 0.02-2,000 μM; decreases melanin content to 20.9% of control in B16 murine melanoma cells at 25 μg/ml; inhibits TPA-induced activation of EBV-EA in Raji cells (IC50 = 499 mol ratio/32 pmol TPA)