Learn More
ATCC Quantitative Synthetic Norovirus G1 (I) RNA (ATCC VR-3234SD™)
Supplier:  ATCC VR3234SD
Encompass
Quantitative Synthetic Norovirus G1 (I) RNA (ATCC VR-3234SD™) is a quantitative molecular standard containing fragments from the RNA-dependent RNA polymerase and VP1 (ORF1-ORF2 junction) regions. Manufactured under ISO 13485 guidance, this product can be used for assay development, qPCR calibration, and foodborne pathogen detection.
Catalog No. 50-238-2741