Learn More
ATCC Quantitative Synthetic Norovirus G2 (II) RNA (ATCC VR-3235SD™)
Catalog No. 502381927
Encompass
Synthetic Norovirus GII RNA (ATCC VR-3235SD™) is a quantitative molecular standard containing fragments from the RNA-dependent RNA polymerase, VP1 (ORF1-ORF2 junction), and VP2 regions. Used for qPCR assay development, validation, and foodborne pathogen detection. Manufactured under ISO 13485.
Catalog No. 50-238-1927 Supplier ATCC Supplier No. VR3235SD
Synthetic Norovirus GII RNA (ATCC VR-3235SD™) is a quantitative molecular standard containing fragments from the RNA-dependent RNA polymerase, VP1 (ORF1-ORF2 junction), and VP2 regions. Used for qPCR assay development, validation, and foodborne pathogen detection. Manufactured under ISO 13485.