Learn More
Cayman Chemical Ombuin, Purity: ≥98%, CAS: 529-40-8, 5 mg
SDP
Catalog No. 502943513
Encompass_Preferred
A flavonoid with diverse biological activities; an antagonist of M1 mAChRs (Ki = 42 µM in CHO-K1 cells expressing the human receptor); cytotoxic to HepG2 cancer cells (IC50 = 1.5 µg/ml); reduces blood urea and glucose levels, urinary volume and protein levels, serum creatinine levels, renal fibrosis levels, and body weight in a rat model of STZ-induced diabetic nephropathy at 40 mg/kg per day
Catalog No. 50-294-3513 Supplier Cayman Chemical Supplier No. 402175 MG
A flavonoid with diverse biological activities; an antagonist of M1 mAChRs (Ki = 42 µM in CHO-K1 cells expressing the human receptor); cytotoxic to HepG2 cancer cells (IC50 = 1.5 µg/ml); reduces blood urea and glucose levels, urinary volume and protein levels, serum creatinine levels, renal fibrosis levels, and body weight in a rat model of STZ-induced diabetic nephropathy at 40 mg/kg per day