Learn More
Utak SMx Oral Fluid 1x100mL
SDP
Catalog No. NC1561552
Encompass_Preferred
Our Specialty Matrix (SMx) Oral Fluid is a synthetic product designed specifically to mimic real human oral fluid while eliminating unwanted endogenous analytes. SMx Oral Fluid is an alternative to buffer solutions that are not representative of human saliva.SMx Oral Fluid can be used on any testing platform, including LCMS, LC-MS/MS, GCMS, and Time of Flight (TOF).Product Contains: SMx Oral FluidMatrix: Oral FluidUnit Size: 1x100mLForm: FrozenShelf Life: 24 monthsStorage: Store fresh frozen control material at or below -10℃ (14℉).
This item is not returnable. View return policy
Catalog No. NC1561552 Supplier Utak Supplier No. 35400SMXOF(F)
Our Specialty Matrix (SMx) Oral Fluid is a synthetic product designed specifically to mimic real human oral fluid while eliminating unwanted endogenous analytes. SMx Oral Fluid is an alternative to buffer solutions that are not representative of human saliva.SMx Oral Fluid can be used on any testing platform, including LCMS, LC-MS/MS, GCMS, and Time of Flight (TOF).Product Contains: SMx Oral FluidMatrix: Oral FluidUnit Size: 1x100mLForm: FrozenShelf Life: 24 monthsStorage: Store fresh frozen control material at or below -10℃ (14℉).