Learn More
Cayman Chemical Ouabain-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502942334
Encompass_Preferred
An internal standard for the quantification of ouabain by GC- or LC-MS
Catalog No. 50-294-2334 Supplier Cayman Chemical Supplier No. 401681 MG
An internal standard for the quantification of ouabain by GC- or LC-MS