Learn More
ATCC Synthetic Plasmodium; malariae DNA
Catalog No. 502382640
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes the 18S rRNA gene, untranslated region (UTR), cyclooxygenase 1 and 3 (Cox1 & Cox3) and Cytochrome B (Cytb).
Catalog No. 50-238-2640 Supplier ATCC Supplier No. PRA3001SD
 Restricted Item
Add to Cart
Add to Cart
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes the 18S rRNA gene, untranslated region (UTR), cyclooxygenase 1 and 3 (Cox1 & Cox3) and Cytochrome B (Cytb).