Learn More
ATCC Synthetic Plasmodium; vivax DNA
Catalog No. 502383823
Encompass
Quantitative Synthetic Plasmodium vivax DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes fragments from 18s rRNA, mitochondrial DNA, cox3, cox1, cytB, and aspartic protease PM4 regions.
Catalog No. 50-238-3823 Supplier ATCC Supplier No. PRA3004SD
Quantitative Synthetic Plasmodium vivax DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes fragments from 18s rRNA, mitochondrial DNA, cox3, cox1, cytB, and aspartic protease PM4 regions.