Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Synthetic Rubber Tygon Hose
SDP

Catalog No. 05405073 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Application):
Vacuum Connection
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
05-405-073 Vacuum Connection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-405-073 Supplier BUCHI Supplier No. 11063244
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic rubber tygon tubing features an inner diameter of 6 mm, and an outer diameter of 13 mm.

  • For use with vacuum
  • Inner Diameter: 6 mm
  • Outer Diameter: 13 mm

Specifications

Material Synthetic Rubber
Type Vacuum Hose
For Use With (Application) Vacuum Connection
For Use With (Equipment) V-100, V-300, V-600, V-700, V-710, R-100, R-210, R-215, R-300, R II
Diameter (Metric) 6 mm (Inner), 13 mm (Outer)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.