Learn More
ATCC Synthetic Sapovirus; RNA
Catalog No. 502382745
Encompass
Quantitative synthetic RNA for Sapovirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the RNA-dependent RNA polymerase, VP1, and polyprotein regions.
Catalog No. 50-238-2745 Supplier ATCC Supplier No. VR3237SD
Quantitative synthetic RNA for Sapovirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the RNA-dependent RNA polymerase, VP1, and polyprotein regions.