Learn More
Cayman Chemical SynthetIc ScoulerIn 1mg
SDP
Catalog No. 502911488
Encompass_Preferred
A benzylisoquinoline alkaloid with diverse biological activities; selectively binds to the dopamine D1 and D2 receptors (Kis = 22 and 214 nM, respectively) over 5-HT1A and 5-HT2A receptor subtypes in HEK293 cells at 10 µM; inhibits proliferation in a panel of leukemia cell lines (IC50s = 2.7-6.5 µM); inhibits methamphetamine-induced conditioned place preference in mice at 5 mg/kg
Catalog No. 50-291-1488 Supplier Cayman Chemical Supplier No. 351401 MG
A benzylisoquinoline alkaloid with diverse biological activities; selectively binds to the dopamine D1 and D2 receptors (Kis = 22 and 214 nM, respectively) over 5-HT1A and 5-HT2A receptor subtypes in HEK293 cells at 10 µM; inhibits proliferation in a panel of leukemia cell lines (IC50s = 2.7-6.5 µM); inhibits methamphetamine-induced conditioned place preference in mice at 5 mg/kg