Learn More
Cayman Chemical Tapinarof, Purity: ≥98%, CAS: 79338-84-4, 10 mg
SDP
Catalog No. 502912899
Encompass_Preferred
A stilbene with diverse biological activities; active against C. tropicales, C. neoformans, A. fumigatus, A. flavus, and B. cinerea (MICs = 25, 12, 12, 25, 12 µg/ml, respectively); induces nuclear translocation of AhR in HaCaT keratinocytes (EC50 = 0.16 nM); induces expression of the AhR target gene CYP1A1 in HaCaT keratinocytes at 1µM; reduces activated immune cell-induced mRNA expression of the gene encoding IL-17a in isolated human keratinocytes at 10 µM
Catalog No. 50-291-2899 Supplier Cayman Chemical Supplier No. 3652710 MG
A stilbene with diverse biological activities; active against C. tropicales, C. neoformans, A. fumigatus, A. flavus, and B. cinerea (MICs = 25, 12, 12, 25, 12 µg/ml, respectively); induces nuclear translocation of AhR in HaCaT keratinocytes (EC50 = 0.16 nM); induces expression of the AhR target gene CYP1A1 in HaCaT keratinocytes at 1µM; reduces activated immune cell-induced mRNA expression of the gene encoding IL-17a in isolated human keratinocytes at 10 µM