Learn More
Cayman Chemical SynthetIc Thujopsen 1mg
SDP
Catalog No. 502909997
Encompass_Preferred
A sesquiterpene with diverse biological activities; inhibits Na+/K+-ATPase (IC50 = 25.9 UG/ml) and CYP2B6 (Ki = 0.8 µM); active against S. aureus, M. luteus, and S. typhimurium (MICs = 25-50 UG/ml); inhibits antigen-induced β-hexosaminidase release from IgE-sensitized RBL-2H3 mast cells (IC50 = 25.1 µM); cytotoxic to A549 cells (LC50 = 35.27 UG/ml); induces mortality in the mites D. farniae and T. putrescentiae (LC50s = 9.82 and 10.92 UG/cm2, respectively)
Catalog No. 50-290-9997 Supplier Cayman Chemical Supplier No. 352701 MG
A sesquiterpene with diverse biological activities; inhibits Na+/K+-ATPase (IC50 = 25.9 UG/ml) and CYP2B6 (Ki = 0.8 µM); active against S. aureus, M. luteus, and S. typhimurium (MICs = 25-50 UG/ml); inhibits antigen-induced β-hexosaminidase release from IgE-sensitized RBL-2H3 mast cells (IC50 = 25.1 µM); cytotoxic to A549 cells (LC50 = 35.27 UG/ml); induces mortality in the mites D. farniae and T. putrescentiae (LC50s = 9.82 and 10.92 UG/cm2, respectively)