Learn More
Cayman Chemical Thymol, Purity: ≥95%, CAS: 89-83-8, 100 mg
SDP
Catalog No. 502948039
Encompass_Preferred
A terpenoid with diverse biological activities; activates TRPV3 currents in HEK293 cells expressing mouse or human TRPV3 at 500 µM; active against a variety of plant pathogenic fungi (MICs = 100-300 µg/ml); active against E. coli and S. aureus (MIC = 500 µg/ml for both); inhibits lipid peroxidation in ox brain phospholipid liposomes by 45-82% at 0.067-0.664 mM; reduces inflammatory cell infiltration into the airway, decreases the levels of IL-4, IL-5, and IL-13 in BALF, and prevents airway hyperresponsiveness in a mouse model of ovalbumin-induced allergic asthma at 16 mg/kg
Catalog No. 50-294-8039 Supplier Cayman Chemical Supplier No. 34496100 MG
A terpenoid with diverse biological activities; activates TRPV3 currents in HEK293 cells expressing mouse or human TRPV3 at 500 µM; active against a variety of plant pathogenic fungi (MICs = 100-300 µg/ml); active against E. coli and S. aureus (MIC = 500 µg/ml for both); inhibits lipid peroxidation in ox brain phospholipid liposomes by 45-82% at 0.067-0.664 mM; reduces inflammatory cell infiltration into the airway, decreases the levels of IL-4, IL-5, and IL-13 in BALF, and prevents airway hyperresponsiveness in a mouse model of ovalbumin-induced allergic asthma at 16 mg/kg