Learn More
Cayman Chemical Tubercidin, Purity: ≥98%, CAS: 69-33-0, 5 mg
SDP
Catalog No. 502910848
Encompass_Preferred
A nucleoside analog with diverse biological activities; active against S. faecalis (IC50 = 0.02 µM); cytotoxic to P388 and A549 cells (IC50s = 0.008 and 0.019 µg/ml, respectively); reduces infection of Calu-3 cells by SARS-CoV-2 (EC50 = 0.05 µM) but is toxic to A549-ACE2 and Vero cells; increases survival in a mouse model of schistosomiasis when used at 5 mg/kg in combination with NBMPR-P but is toxic and induces mortality when administered alone at the same dose for four days
Catalog No. 50-291-0848 Supplier Cayman Chemical Supplier No. 358345 MG
A nucleoside analog with diverse biological activities; active against S. faecalis (IC50 = 0.02 µM); cytotoxic to P388 and A549 cells (IC50s = 0.008 and 0.019 µg/ml, respectively); reduces infection of Calu-3 cells by SARS-CoV-2 (EC50 = 0.05 µM) but is toxic to A549-ACE2 and Vero cells; increases survival in a mouse model of schistosomiasis when used at 5 mg/kg in combination with NBMPR-P but is toxic and induces mortality when administered alone at the same dose for four days