Learn More
Cayman Chemical SynthetIc Uvaol 10mg
SDP
Catalog No. 502950090
Encompass_Preferred
A pentacyclic triterpenoid; induces cell cycle arrest at the G0/G1 phase and inhibits proliferation of MCF-7 human breast cancer cells in a concentration-dependent manner; prevents hydrogen peroxide-induced DNA damage in the same cells; induces relaxation of phenylephrine- or potassium chloride-precontracted aortic rings isolated from spontaneously hypertensive rats at 100 µM; inhibits ovalbumin-induced BALF eosinophil infiltration in an ovalbumin-sensitized mouse model of allergic asthma
Catalog No. 50-295-0090 Supplier Cayman Chemical Supplier No. 3391510 MG
A pentacyclic triterpenoid; induces cell cycle arrest at the G0/G1 phase and inhibits proliferation of MCF-7 human breast cancer cells in a concentration-dependent manner; prevents hydrogen peroxide-induced DNA damage in the same cells; induces relaxation of phenylephrine- or potassium chloride-precontracted aortic rings isolated from spontaneously hypertensive rats at 100 µM; inhibits ovalbumin-induced BALF eosinophil infiltration in an ovalbumin-sensitized mouse model of allergic asthma