Learn More
Waters Corp Waters Corporation Synthetic Vacuum Pump Oil, 1 L, For Mass Spectrometers
Catalog No. 502224485 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement synthetic vacuum pump oil is specifically designed for use with Leybold Pumps on a range of mass spectrometers, ensuring optimal performance and longevity of your equipment.
  • Synthetic type of oil
  • Volume: 1 L
  • Used with Leybold Pumps
  • Compatible with mass spectrometers
  • Annual replacement schedule
Catalog No. 50-222-4485 Supplier Waters Corp Supplier No. 700012074
This replacement synthetic vacuum pump oil is specifically designed for use with Leybold Pumps on a range of mass spectrometers, ensuring optimal performance and longevity of your equipment.
  • Synthetic type of oil
  • Volume: 1 L
  • Used with Leybold Pumps
  • Compatible with mass spectrometers
  • Annual replacement schedule