Learn More
Cayman Chemical Vitamin K3, Purity: ≥95%, CAS: 58-27-5, 100 g
SDP
Catalog No. 502972153
Encompass_Preferred
A precursor to Vitamin K2 that is capable of both redox cycling and arylating nucleophilic substrates by Michael addition; used as an antihemorrhagic agent and to inhibit the proliferation of various cancer cells
Catalog No. 50-297-2153 Supplier Cayman Chemical Supplier No. 15950100 G
A precursor to Vitamin K2 that is capable of both redox cycling and arylating nucleophilic substrates by Michael addition; used as an antihemorrhagic agent and to inhibit the proliferation of various cancer cells