Learn More
Cayman Chemical XAV939, Purity: ≥98%, CAS: 284028-89-3, 10 mg
SDP
Catalog No. 502955654
Encompass_Preferred
A small molecule inhibitor of tankyrase 1 and 2 (IC50 = 11 and 4 nM, respectively); increases the protein levels of the axin-GSK3β complex and promotes the degradation of β-catenin; inhibits colony formation of APC-deficient colorectal cancer cells at 0.33 µM
Catalog No. 50-295-5654 Supplier Cayman Chemical Supplier No. 1359610 MG
A small molecule inhibitor of tankyrase 1 and 2 (IC50 = 11 and 4 nM, respectively); increases the protein levels of the axin-GSK3β complex and promotes the degradation of β-catenin; inhibits colony formation of APC-deficient colorectal cancer cells at 0.33 µM