Learn More
ATCC Synthetic Zika; virus RNA
Catalog No. 502382150
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Quantitative synthetic RNA for Zika virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the Membrane glycoprotein precursor M, Envelope, NS1, NS2B, NS3, NS4B, and NS5 regions.
Catalog No. 50-238-2150 Supplier ATCC Supplier No. VR3252SD
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Quantitative synthetic RNA for Zika virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the Membrane glycoprotein precursor M, Envelope, NS1, NS2B, NS3, NS4B, and NS5 regions.