Learn More
eMolecules​ 1314538-55-0 | Potassium (((tert-butoxycarbonyl)amino)methyl)trifluoroborate | Synthonix | MFCD19686142 | 237.070 | C6H12BF3KNO2 | 95.000 | [K+].CC(C)(C)OC(=O)NC[B-](F)(F)F | 100mg | 649428401
Catalog No. 5000126742 Shop All eMolecules Inc. Products
Encompass_Preferred
Potassium (((tert-butoxycarbonyl)amino)methyl)trifluoroborate | Synthonix | 1314538-55-0 | MFCD19686142 | 237.070 | C6H12BF3KNO2 | 95.000 | [K+].CC(C)(C)OC(=O)NC[B-](F)(F)F | 100mg | 649428401
Catalog No. 50-001-26742 Supplier eMolecules​ Supplier No. 649428401
Potassium (((tert-butoxycarbonyl)amino)methyl)trifluoroborate | Synthonix | 1314538-55-0 | MFCD19686142 | 237.070 | C6H12BF3KNO2 | 95.000 | [K+].CC(C)(C)OC(=O)NC[B-](F)(F)F | 100mg | 649428401