Description
- Synthware combination adapting connector with a top spherical socket joint and a lower standard taper inner joint.
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass (3.3) |
| Product Type | Adapter with Ball Socket |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 Bottom, 35/20 Top |