Learn More
Synthware™ Bushing Adapters
GREENER_CHOICESDP
Catalog No. 31500505 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
19/22 Inner Bottom, 14/20 Outer Top
19/38 Inner Bottom, 14/35 Outer Top
24/40 Inner Bottom, 14/35 Outer Top
24/40 Inner Bottom, 19/38 Outer Top
34/45 Inner Bottom, 24/40 Outer Top
45/50 Inner Bottom, 24/40 Outer Top
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-505 45/50 Inner Bottom, 24/40 Outer Top
31-500-500 19/22 Inner Bottom, 14/20 Outer Top
31-500-501 19/38 Inner Bottom, 14/35 Outer Top
31-500-502 24/40 Inner Bottom, 14/35 Outer Top
31-500-503 24/40 Inner Bottom, 19/38 Outer Top
31-500-504 34/45 Inner Bottom, 24/40 Outer Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-500-505 Supplier Synthware™ Supplier No. A792445
Add to Cart
Add to Cart
For compact assemblies of dissimilar joints
  • Synthware bushing adapters allow for compact assemblies of dissimilar standard taper joints.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Bushing adapter
Quantity 1
Standard Taper Joint No. 45/50 Inner Bottom, 24/40 Outer Top