Description
- Synthware Claisen Adapter.
- Provides dual entry into a variety of flasks.
- Adapter has standard taper outer joints at top and on side and inner joint on bottom.
- All joints are the same size.
Specifications
Specifications
| Includes | Two-holed nylon compression caps, two Viton O-rings and two PTFE-lined silicone septa |
| Material | Borosilicate Glass (3.3) |
| Product Type | Claisen Adpater |
| Standard Taper Joint No. | 14/10 |