Learn More
Synthware™ Connecting Adapter
GREENER_CHOICESDP
Catalog No. 31500587 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
24/40
Catalog No. Standard Taper Joint No.
31-500-587 24/40
1 options
This item is not returnable. View return policy
Catalog No. 31-500-587 Supplier Synthware™ Supplier No. A192424
Add to Cart
Add to Cart
This item is not returnable. View return policy
Converts standard vessels to airless applications.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Connecting adapter
Quantity 1
Standard Taper Joint No. 24/40