Description
- Synthware connecting adapter is a 3-way adapter used for connecting three vessels
- It has two top outer joints and a lower inner joint, all being the same size
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass (3.3) |
| Product Type | Connecting adapter |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 |