Description
- Sidearm has standard taper inner joint
- Vertical tube has top outer and lower inner joint all being the same size
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Product Type | Distillation Adapter |
| Standard Taper Joint No. | 24/40, 10/18 Thermometer |