Learn More
Synthware™ Distillation Adapter, Bent, 105°
GREENER_CHOICESDP
Catalog No. 31500600 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
24/40
Catalog No. Standard Taper Joint No.
31-500-600 24/40
1 options
Catalog No. 31-500-600 Supplier Synthware™ Supplier No. A412440
Add to Cart
Add to Cart
To effectively clean the rotavapor condenser
  • With top outer and lower inner standard taper joints
  • Both joints are of same size
Specifications
Material Borosilicate Glass (3.3)
Product Type Distillation adapter
Standard Taper Joint No. 24/40