Learn More
Synthware™ Distillation Connecting Adapters, Microscale
GREENER_CHOICESDP
Catalog No. 31502093 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/10
Catalog No. Standard Taper Joint No.
31-502-093 14/10
1 options
Catalog No. 31-502-093 Supplier Synthware™ Supplier No. A351410
Add to Cart
Add to Cart
Distillation adapter with one holed Nylon™ compression cap and one Viton™ O-ring. For microscale applications
  • Supplied with one holed compression cap and one Viton™ O-ring
  • Joint: 14/10
Specifications
Includes One holed compression cap and one Viton™ O-ring.
Material Borosilicate Glass (3.3)
Product Type Distillation Connecting Adapter
Standard Taper Joint No. 14/10