Learn More
Synthware™ Distillation Adapters
GREENER_CHOICESDP
Catalog No. 31500679 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-679 24/40
31-500-677 14/20
31-500-678 19/22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-679 Supplier Synthware™ Supplier No. A902440
Add to Cart
Add to Cart
With threaded thermometer port on the side arm
  • Accommodates 6 to 7.7mm O.D. plain stem thermometer
  • Sidearm with standard taper inner joint with drip tip
  • Vertical tube has top outer and lower inner joint
  • Supplied with open top cap and Viton™ O-ring
Specifications
Includes Open top cap and Viton™ O-ring
Material Borosilicate Glass (3.3)
Product Type Distillation adapter
Standard Taper Joint No. 24/40