Description
- Synthware distillation adapter with sealed in drip tip
- Adapter is bent at 105 degrees
- Standard taper top outer joint and lower inner joints
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass (3.3) |
| Product Type | Distilling Adapter, Bent, 105 Degree |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 Bottom, 24/40 Top |