Description
- Thermometer stem and bulb serves as column packing
- Lower hose connection for vacuum
- Upper hose connections for cooling lines
- Used with 25mm immersion thermometer
Specifications
Specifications
| Type | Distillation Head |
| Standard Taper Joint | Thermometer Joint 10/18 |
| Joint Type | 24/40 joint |