Learn More
Synthware™ Distilling Head, Short Path, For Volatile Distillants
GREENER_CHOICESDP
Catalog No. 31501026 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
Joint 24/40
Catalog No. Standard Taper Joint
31-501-026 Joint 24/40
1 options
Catalog No. 31-501-026 Supplier Synthware™ Supplier No. D541024
Add to Cart
Add to Cart
Distilling Head, Short Path, For Volatile Distillants with close coupled condenser and vacuum connection to offer an extremely short condensate travel path.
  • Combination of cold coil and Leibig.
  • Thermometer stem and bulb serves as column packing.
  • Lower hose connection for vacuum; upper hose connections for cooling lines.
  • Used with 50 mm immersion thermometer.
  • Vigreux indentations improve efficiency.
Specifications
Type Distillation Head
Standard Taper Joint Joint 24/40
Joint Type Thermometer Joint 10/18