Description
- Short path distilling head with close coupled condenser and vacuum connection to offer extremely short condensate travel path
- Condenser is combination of cold coil and Leibig
- Thermometer stem and bulb serves as column packing
- Lower hose connection for vacuum; upper hose connections for cooling lines
- Used with 50mm immersion thermometer
Specifications
Specifications
| Type | Distillation Head |
| Standard Taper Joint | Joint 24/40 |
| Joint Type | Thermometer Joint 10/18 |