Learn More
Synthware™ Distilling Head, Short Path, Jacketed
GREENER_CHOICESDP
Catalog No. 31501021 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20 Joint
Catalog No. Standard Taper Joint
31-501-021 14/20 Joint
1 options
Catalog No. 31-501-021 Supplier Synthware™ Supplier No. D121014
Add to Cart
Add to Cart
Short path distilling head has a close coupled condenser and vacuum connection to offer an extremely short condensate travel path
  • Thermometer stem and bulb serve as column packing
  • Lower hose connection for vacuum
  • Upper hose connections for cooling lines
  • Used with 25mm immersion thermometer
  • Vacuum jacketed design for more efficiency
Specifications
Type Distillation Head
Standard Taper Joint 14/20 Joint
Joint Type Thermometer Joint: 10/18