Learn More
Synthware™ Distilling Trap Adapters
GREENER_CHOICESDP
Catalog No. 31500609 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
Upper: 24/40, Lower: 14/20
Upper: 24/40, Lower: 19/22
Upper: 24/40, Lower: 24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-500-609 Upper: 24/40, Lower: 14/20
31-500-610 Upper: 24/40, Lower: 19/22
31-500-611 Upper: 24/40, Lower: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-609 Supplier Synthware™ Supplier No. A262414
Add to Cart
Add to Cart
Placed above flask to prevent carry-over of reactions which have a tendency to foam
Synthware Kjeldahl distilling trap adapter. Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam. With top outer and lower inner standard taper joints.
Specifications
Standard Taper Joint Upper: 24/40, Lower: 14/20
Material Borosilicate Glass (3.3)
Capacity (Metric) 100 mL
Quantity 1
For Use With (Application) Organic Synthesis
Product Type Distilling Trap Adapter