Description
- Synthware Kjeldahl distilling trap adapter
- Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam
- With top outer and lower inner standard taper joints
Specifications
Specifications
| Standard Taper Joint | Upper: 24/40, Lower: 24/40 |
| Material | Borosilicate Glass (3.3) |
| Capacity (Metric) | 100 mL |
| Quantity | 1 |
| For Use With (Application) | Organic Synthesis |
| Product Type | Distilling Trap Adapter |