Description
- Synthware drying tube angled at 75 degrees
- Angled to avoid dessicant contact with reactions
- With standard taper inner joint
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass (3.3) |
| Product Type | Drying Tube Adapter, Angled at 75° |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 |