Description
- Synthware straight drying tube
- With top outer and lower inner standard taper joints of the same size
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass (3.3) |
| Product Type | Drying Tube Adapter |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 |