Description
- Synthware vacuum adapter with a serrated hose connection turned 90 degrees to the standard taper outer joint
- The glass stopcock has a 2 mm bore and is used to regulate the flow of gasses or liquids into reaction systems
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Includes | Glass Stopcock and Outer Joint |
| Material | Borosilicate Glass (3.3) |
| Product Type | Flow Control Adapter |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 |