Learn More
Synthware™ Flow Control Adapters with Glass Stopcock and Outer Joint
GREENER_CHOICESDP
Catalog No. p-6629007 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-569 14/20
31-500-570 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-569 Supplier Synthware™ Supplier No. A691420
Only null left
Add to Cart
Add to Cart
For regulating the input of gases or liquids into reaction systems
Synthware vacuum adapter with a serrated hose connection turned 90 degrees to the standard taper outer joint. The glass stopcock has a 2 mm bore and is used to regulate the flow of gasses or liquids into reaction systems.
Specifications
For Use With (Application) Organic Synthesis
Item Description Flow Control Adapter with glass stopcock and outer joint
Material Borosilicate Glass (3.3)
Product Type Flow Control Adapter
Quantity 1
Standard Taper Joint No. 14/20