Learn More
Synthware™ Flow Control Adapters with PTFE Stopcock
GREENER_CHOICESDP
Catalog No. 31500558 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-558 19/22
31-500-559 24/40
31-500-557 14/20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-558 Supplier Synthware™ Supplier No. A701922
Add to Cart
Add to Cart
Used to regulate the flow of gases or liquids into reaction systems
  • With standard taper inner joint
  • 2mm stopcock bore
Specifications
Includes PTFE 2mm stopcock
Material PTFE
Product Type Flow control adapter
Standard Taper Joint No. 19/22