Learn More
Synthware™ Friedrichs Condenser
GREENER_CHOICESDP
Catalog No. 31500736 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint Bottom:
24/40
Catalog No. Standard Taper Joint Bottom
31-500-736 24/40
1 options
Catalog No. 31-500-736 Supplier Synthware™ Supplier No. C232440
Add to Cart
Add to Cart
Spiral inner tube with long vapor path providing good heat transfer and anti-flooding characteristics
  • Lower inner joint has a drip tip
Specifications
For Use With (Equipment) Distillation Head
Product Type Friedrichs Condenser
Standard Taper Joint Bottom 24/40
Diameter (Metric) Outer 10 mm hose connection
Height (Metric) 330 mm