Learn More
Synthware™ Gas Inlet/Outlet Adapter, Microscale
GREENER_CHOICESDP
Catalog No. 31502089 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/10
Catalog No. Standard Taper Joint No.
31-502-089 14/10
1 options
Catalog No. 31-502-089 Supplier Synthware™ Supplier No. A301410
Add to Cart
Add to Cart
Gas inlet/outlet adapter with 14/10 joint. For microscale applications
  • Joint 14/10
  • Hose Connection 8
Specifications
Material Borosilicate Glass (3.3)
Product Type Inlet/Oulet Adapter
Standard Taper Joint No. 14/10