Learn More
Synthware™ Inlet Adapters with Fritted Disc
GREENER_CHOICESDP
Catalog No. 31500545 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
Height (Metric):
65mm
67mm
95mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No. Height (Metric)
31-500-545 14/20 65mm
31-500-546 19/22 67mm
31-500-547 24/40 95mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-545 Supplier Synthware™ Supplier No. A831420
Add to Cart
Add to Cart
With extra coarse porosity fritted disc to prevent solid from being introduced into reaction system during liquid transfer
Synthware inlet adapter with an extra coarse porosity fritted disc between the serrated hose connection and the standard taper inner joint. The disc prevents solids from being introduced into the reaction system during liquid transfer.
Specifications
For Use With (Application) Organic Synthesis
Height (Metric) 65mm
Material Borosilicate Glass (3.3)
Product Type Inlet Adapter with Fritted Disc
Quantity 1
Standard Taper Joint No. 14/20