Description
- Synthware inlet adapter with an extra coarse porosity fritted disc between the serrated hose connection and the standard taper inner joint
- The disc prevents solids from being introduced into the reaction system during liquid transfer
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Height (Metric) | 95mm |
| Material | Borosilicate Glass (3.3) |
| Product Type | Inlet Adapter with Fritted Disc |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 |