Description
With a compression cap and Viton™ O-ring to form a vacuum tight seal
- To use as syringe port, replace O-ring with PTFE-lined septum
- Cap accommodates 5.0-7.5mm O.D. thermometer
- Supplied with one Nylon™ cap and one Vilton™ O-ring
Specifications
Specifications
| Includes | Compression Nylon cap and Viton O-ring |
| Material | Borosilicate Glass (3.3) |
| Product Type | Thermometer Adapter |
| Standard Taper Joint No. | 14/10 |