Description
- Cap accommodates 5.0to 7.5mm O.D. thermometer
- Supplied with one Nylon™ cap and one Viton™O-ring
Specifications
Specifications
| Material | Borosilicate Glass |
| Product Type | Inlet or Thermometer Adapter |
| Standard Taper Joint No. | 24/40 |