Learn More
Synthware™ Reducing Adapters
GREENER_CHOICESDP
Catalog No. 31500509 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
19/22 Bottom, 14/20 Top
24/40 Bottom, 10/30 Top
24/40 Bottom, 14/20 Top
24/40 Bottom, 19/22 Top
24/40 Bottom, 34/45 Top
29/42 Bottom, 24/40 Top
45/50 Bottom, 24/40 Top
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-509 24/40 Bottom, 19/22 Top
31-500-506 24/40 Bottom, 10/30 Top
31-500-507 19/22 Bottom, 14/20 Top
31-500-508 24/40 Bottom, 14/20 Top
31-500-510 29/42 Bottom, 24/40 Top
31-500-511 24/40 Bottom, 34/45 Top
31-500-512 45/50 Bottom, 24/40 Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-500-509 Supplier Synthware™ Supplier No. A201924
Add to Cart
Edge
Add to Cart
With standard taper joints at top and bottom
  • Synthware reducing adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is larger in size than the top outer joint.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Reducing adapter
Quantity 1
Standard Taper Joint No. 24/40 Bottom, 19/22 Top